Learn More
VACUUBRAND™ Metal Vacuum Tubing With Small Flanges KF
A durable connection option for the laboratory that still provides flexibility.
Brand:  VACUUBRAND™ 20673338
 View more versions of this product
Product Code. 10046679
  • 334.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from stainless steel
  • Available in a range of sizes and lengths
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuum Tubing
40 mm
100 cm
DN 40
Single bending radius of 65
Metal
1 Each