Learn More
VACUUBRAND™ Metal Vacuum Tubing With Small Flanges KF
A durable connection option for the laboratory that still provides flexibility.
Brand:  VACUUBRAND™ 20673328
 View more versions of this product
Product Code. 10340742
  • 299.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from stainless steel
  • Available in a range of sizes and lengths
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuum Tubing
40 mm
75 cm
DN 40
Single bending radius of 65
Metal
1 Each