Learn More
Brady™ Metal Wall Lock Box
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Metal Wall Lock Box is made of durable, powder-coated steel with key slot on top. Includes clear Lexan window that slides out when locks are removed.
Brand:  Brady™ 105714
 View more versions of this product
Product Code. 15611127
  • 126.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
Wall Mount Lock Box
Powder-coated Steel
GROUP LOCK BOX
(7) key hooks
1 Ea.
Yellow on Red
2.5 x 8 x 7 in.
7 in.
2.5 in.
8 in.