Learn More
3M™ Metaliks™ Safety Glasses
3M™ Metaliks™ Safety Glasses have a polished metal frame with a sleek 10-base de-centered twin lens for optimum eye coverage and optimal field of vision.
299.68 € - 351.00 €
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
---|---|
Certifications/Compliance | CE Approved, EN 166: 2001 |
Product Type | Safety glasses |
Lens Material | Polycarbonate |
Frame Material | Metal |
Description
- Comfortable spatula temples with soft tips for a perfect fit
- Soft adjustable nose pads to custom fit virtually all face shapes and sizes
- Optical class 1 for maximum clarity
- Anti-Scratch and Anti-Fog coating for increased durability and improved vision
- Soft-touch temple tips provide additional comfort
Specifications
Anti-fog, Scratch-resistant | |
Safety glasses | |
Metal |
CE Approved, EN 166: 2001 | |
Polycarbonate |