missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Metalware Range™ Stainless Steel Square Sterilizing Pipet Cans

Reduces the risk of damage to pipets during transportation and sterilisation. Nickel Electro™ Metalware Range™ Stainless Steel Square Sterilization Cans withstand dry heat temperatures of up to 180°C and can be steam sterilized at 120°C.

Brand:  Nickel Electro™ 6700/SS

 View more versions of this product

Product Code. 12946986

  • 55.10 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Popular square pipet cans with permanent cast silicone rubber pads in lids and bases reduce the risk of damage to pipets during transportation and sterilisation.
  • Manufactured from stainless steel.
  • Can be steam sterilized at 120°C.
  • Withstands dry heat temperatures of up to 180°C.
  • Square section design allows easy stacking for efficient use of space and prevents cans from rolling off benchtop.
Specifications

Specifications

Autoclavable
Square Pipet Can
70 x 70 x 210 mm
70 mm
1 kg
Stainless Steel
Permanent cast silicone rubber pads inlids and bottoms
70 mm
210 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.