missing translation for 'onlineSavingsMsg'
Learn More

Metaphosphoric acid, ACS, 33.5-36.5%, bal. NaPO{3} (Stablizer), Thermo Scientific Chemicals

35.28 € - 193.97 €

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357578
View Documents
Thermo Scientific Alfa Aesar
033267.09
10 g
35.28 €
10g
Please to purchase this item. Need a web account? Register with us today!
11367578
View Documents
Thermo Scientific Alfa Aesar
033267.22
100 g
52.26 €
100g
Please to purchase this item. Need a web account? Register with us today!
11377578
View Documents
Thermo Scientific Alfa Aesar
033267.36
500 g
193.97 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Metaphosphoric acid is used as a dehydrating agent. Its derivative, sodium hexametaphosphate is used as a sequestrant and a food additive. It is also used as phosphorylating agent, analytical reagent and in dental cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37267-86-0
79.979
UEZVMMHDMIWARA-UHFFFAOYSA-N
3084658
HO3P
MFCD00011351
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
OP(=O)=O

Specifications

White crystalline
37267-86-0
Hygroscopic
Crystalline
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
UEZVMMHDMIWARA-UHFFFAOYSA-N
79.979
79.98
Metaphosphoric acid
White
2.2 g/cm3 at 20&de g;C
HO3P
MFCD00011351
Decomposes in water. Soluble in alcohol.
OP(=O)=O
3084658
ACS
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : DOT Class: 8; Packing Group: III

missing translation for 'einecsNumber' : 253-433-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.