Learn More
Metaphosphoric Acid, Extra Pure, SLR, Fisher Chemical™
HO3P, CAS Number-37267-86-0, unii-mtk99r3uv0, x-phosphate-group, m-phosphoric acid, metaphosphoric acid hpo3, meta-phosphoric acid, phosphenic acid, metaphosphoric acid, phosphonate 1-, hydrogenphosphonate, hydrogen phosphonate, 500g, Colorless, UEZVMMHDMIWARA-UHFFFAOYSA-N, 200 deg.C, 11351, 79.979
Brand: Fisher Chemical M/3851/53
308.35 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
HO3P, CAS Number-37267-86-0, unii-mtk99r3uv0, x-phosphate-group, m-phosphoric acid, metaphosphoric acid hpo3, meta-phosphoric acid, phosphenic acid, metaphosphoric acid, phosphonate 1-, hydrogenphosphonate, hydrogen phosphonate, 500g, Colorless, UEZVMMHDMIWARA-UHFFFAOYSA-N, 200 deg.C, 11351, 79.979
Specifications
2 | |
37267-86-0 | |
Plastic powder jar | |
HO3P | |
11351 | |
UEZVMMHDMIWARA-UHFFFAOYSA-N | |
79.979 |
Colorless | |
200°C | |
500 g | |
Solid | |
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
OP(=O)=O | |
3084658 |
Safety and Handling
- Metaphosphoric acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.