missing translation for 'onlineSavingsMsg'
Learn More

Metaphosphoric acid, stabilized, Thermo Scientific Chemicals

Product Code. 10105040
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Metaphosphoric acid, 40 to 46% (HPO3), HO3P, CAS Number-37267-86-0, 50813-16-6, hydrogenphosphonate, metaphosphoric acid, meta-phosphoric acid, hydrogen phosphonate, phosphenic acid, m-phosphoric acid, metaphosphoric acid hpo3, phosphonate 1-, x-phosphate-group, unii-mtk99r3uv0, 500g, 5ppm max.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37267-86-0, 50813-16-6
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Heavy Metals (as Pb) 50ppm max.
Packaging Glass bottle
Quantity 100 g
Linear Formula (HPO3)n
Physical Form Glassy Lump
Fieser 01,665; 16,177
Merck Index 15, 7457
Solubility Information Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol
Formula Weight 79.98
Percent Purity 40 to 46% (HPO3)
Chemical Name or Material Metaphosphoric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Metaphosphoric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.