missing translation for 'onlineSavingsMsg'
Learn More

Methacryloyl chloride, may contain up to ca. 15% cyclic dimer, 97%, stab., Thermo Scientific Chemicals

Product Code. 11466820
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacryloyl chloride is used in the preparation of polymeric materials. It reacts with L-histidine and forms functional monomer. It is also used to prepare monomer 2-methacrylamido-caprolactam, amide monomers by amidation.

Solubility
Miscible with alcohols, ethers and organic solvents. Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-46-7
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.533
MDL Number MFCD00000716
InChI Key VHRYZQNGTZXDNX-UHFFFAOYSA-N
Synonym methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
PubChem CID 13528
IUPAC Name 2-methylprop-2-enoyl chloride
SMILES CC(=C)C(=O)Cl

Specifications

Melting Point -60°C
Density 1.07
Boiling Point 95°C to 96°C
Flash Point 2°C (35°F)
Odor Acrid
Refractive Index 1.444
Quantity 100 g
UN Number UN3488
Beilstein 878175
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohols,ethers and organic solvents. Slightly miscible with water.
Formula Weight 104.54
Percent Purity 97%
Chemical Name or Material Methacryloyl chloride, stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'