Learn More
Methacycline hydrochloride, MedChemExpress
MedChemExpress Methacycline hydrochloride is a tetracycline antibiotic and can inhibits bacterial protein synthesis. Methacycline hydrochloride is a potent epithelial-mesenchymal transition (EMT) inhibitor. Methacycline hydrochloride blocks EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. Methacycline hydrochloride is an antimicrobial and has the potential for pulmonary fibrosis.
Brand: Medchem Express HY-B0449
Description
Methacycline hydrochloride is a tetracycline antibiotic and can inhibits bacterial protein synthesis. Methacycline hydrochloride is a potent epithelial-mesenchymal transition (EMT) inhibitor. Methacycline hydrochloride blocks EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. Methacycline hydrochloride is an antimicrobial and has the potential for pulmonary fibrosis.Specifications
Methacycline hydrochloride | |
Yellow | |
C22H23ClN2O8 | |
DMSO : 50 mg/mL (104.41 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (13.93 mM; Need ultrasonic) | |
478.88 | |
99.53% | |
Solid |
3963-95-9 | |
Research | |
500 mg | |
O[C@@](C(O)=C(C(C1=C2C=CC=C1O)=O)[C@](C2=C)([H])[C@@H]3O)(C(C(C(N)=O)=C4O)=O)[C@]3([H])[C@@H]4N(C)C.Cl | |
478.88 | |
Research | |
COVID-19-immunoregulation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only