missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, silver salt, 99%, Thermo Scientific Chemicals

Product Code. 10127872
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-52-9
Molecular Formula CH3AgO3S
Molecular Weight (g/mol) 202.97
MDL Number MFCD00064795
InChI Key MLKQJVFHEUORBO-UHFFFAOYSA-M
Synonym silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
PubChem CID 159658
IUPAC Name silver;methanesulfonate
SMILES CS(=O)(=O)[O-].[Ag+]

Specifications

Melting Point 252.0°C to 256.0°C
Color Beige to White or Gray
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula CH3SO3Ag
Beilstein 04,4
Solubility Information Solubility in water: soluble.
Formula Weight 202.97
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Methanesulfonic acid, silver salt, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methanesulfonic acid, silver salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanesulfonic acid, silver salt, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.