missing translation for 'onlineSavingsMsg'
Learn More
Methanesulfonic Acid Solution, Honeywell™ Fluka™ Product Code.: 15651570

Methanesulfonic Acid Solution, Honeywell™ Fluka™

Product Code. 15651570 Shop All Honeywell Fluka Products
1 L, Glass Bottle
Click to view available options
Quantity:
10 mL
100 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15651570

Brand: Honeywell-Fluka 178341L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Linear Formula CH3SO3H
UN Number UN3265
Beilstein 1446024
Formula Weight 96.11g/mol
Concentration or Composition (by Analyte or Components) approx. 1M in water
Packaging Glass Bottle
Density 1.039g/mL at 20°C
Chemical Name or Material Methanesulfonic Acid Solution, For ion chromatography
Show More Show Less
compliance-icons
Product Identifier
  • Methanesulfonic acid solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanesulphonic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanesulfonic Acid Solution, Honeywell™ Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.