missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic anhydride, 97%, Thermo Scientific Chemicals

Product Code. 11994561
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7143-01-3
Molecular Formula C2H6O5S2
Molecular Weight (g/mol) 174.18
InChI Key IZDROVVXIHRYMH-UHFFFAOYSA-N

Specifications

Melting Point 69.0°C to 70.0°C
Color Gray to White
Boiling Point 125.0°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 500 g
Synonym cholesteryl chloroformate, cholesterol chloroformate, cholesterol, chloroformate, cholesteryloxycarbonyl chloride, cholest-5-en-3-ol 3.beta.-, carbonochloridate, 1r,3as,3bs,7s,9ar,9bs,11ar-9a,11a-dimethyl-1-2r-6-methylheptan-2-yl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl carbonochloridate, cholesterol chlorocarbonate, chlolesterol chloroformate, cholesterol, chloroformate 8ci
Solubility Information Solubility in water: reacts
SMILES CS(=O)(=O)OS(C)(=O)=O
IUPAC Name methanesulfonyl methanesulfonate
PubChem CID 111262
Formula Weight 174.18
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Methanesulfonic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • Methanesulfonic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.