missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 99.8%, BAKER HPLC ANALYZED, J.T. Baker™
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle, packaged under nitrogen
Description
BAKER ANALYZED™ HPLC grade reagents are a group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for Liquid Chromatography applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.
- Manufactured and tested to assure interference-free separations
- Highly characterized reagents that have unmatched lot-to-lot consistency
- Minimizes recalibration or instrument adjustments
- Solvents are controlled for high assay and low UV absorption, fluorescence, residue and water
- An Actual Lot Analysis is provided on the label
Specifications
Specifications
Melting Point | −98°C |
Absorbance | Max. 0.20 at 225nm, Max. 0.02 at 254nm, Max. 0.01 at 280nm, Max. 0.01 at 350nm |
CAS | 67-56-1 |
Color | Colorless |
Density | 0.79g/cm3 at 20°C |
Boiling Point | 64.7°C |
Quantity | 2.5 L |
Explosion Limit | Lower: 5.5 vol %, Upper: 44 Vol % |
Flash Point | 11°C |
Ignition Point | 455°C |
Show More |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction