missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99.8%, BAKER LC/MS ANALYZED, J.T. Baker™

Product Code. 15514065
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

LC-MS grade methanol for use in Liquid Chromatography and Mass Spectrophotometry, packaged in 2.5L glass bottles.

BAKER ANALYZED™ LC-MS grade reagents are a group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for Liquid Chromatography applications and UV-spectrophotometric analysis.

  • Manufactured and tested to assure interference-free separations
  • Highly characterized reagents that have unmatched lot-to-lot consistency
  • Minimizes recalibration or instrument adjustments
  • Solvents are controlled for high assay and low UV absorption, fluorescence, residue and water
  • An Actual Lot Analysis is provided on the label

Specifications

Melting Point −97.8°C
Absorbance Max. 0.20 at 225nm, Max. 0.02 at 254nm, Max. 0.01 at 280nm
CAS 67-56-1
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 64°C
Quantity 2.5 L
Explosion Limit Lower: 6 vol %, Upper: 36 Vol %
Flash Point 11°C
Ignition Point 240°C
Odor Alcohol
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 1 ppm
Molecular Formula CH4O
MDL Number MFCD00004595
Vapor Pressure 16.9 kPa at 25°C
Water Max. 0.02%
Solubility Information Miscible in ether, benzene, ethanol; 1.000 g/L (in water)
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Percent Purity Min. 99.8%
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'