Learn More
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
25.73 € - 416.07 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10532971
|
Thermo Scientific Acros
176845000 |
500 mL |
25.73 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10317360
|
Thermo Scientific Acros
176840010 |
1 L |
37.79 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10327360
|
Thermo Scientific Acros
176840025 |
2.5 L |
73.16 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10012973
|
Thermo Scientific Acros
176840100 |
10 L |
207.03 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10547183
|
Thermo Scientific Acros
176840250 |
25 L |
416.07 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98.0°C | |
0.7910g/mL | |
12°C | |
Liquid | |
99.9% | |
0.791 | |
0.0003 meq/g max. | |
0.0002 meq/g max. | |
0.55 mPa.s (20°C) | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
Analytical |
67-56-1 | |
64.7°C | |
Glass Bottle | |
1.3280 to 1.3300 | |
0.002% max. | |
CH4O | |
CH3OH | |
MFCD00004595 | |
0.05% max. (K.F.) | |
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents | |
CO | |
32.04 | |
CHEBI:17790 | |
99.9% | |
Methanol |
Sicherheit und Handhabung
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Danger
- Methanol
Signalwort
- Danger
Gefahrenkategorie
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Gefahrenhinweis
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only