missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99+%, extra pure, Thermo Scientific Chemicals

32.36 € - 324.62 €

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10245850
SDS View Documents
Thermo Scientific Acros
177150010
1 L Glass Bottle
32.36 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10674862
SDS View Documents
Thermo Scientific Acros
177150025
2.5 L Glass Bottle
55.17 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10778384
SDS View Documents
Thermo Scientific Acros
177150050
5 L Plastic Drum
94.07 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10792791
SDS View Documents
Thermo Scientific Acros
177150250
25 L Plastic Can
324.62 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, 99+%, extra pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.