Learn More
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Description
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | °C to 98°C |
Color | Colorless |
Density | 0.791 |
Boiling Point | 64.7°C |
Quantity | 4 L |
Flash Point | 11°C (52°F) |
Physical Form | Liquid |
Refractive Index | 1.329 |
Assay Percent Range | >99.8% |
Beilstein | 1098229 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N28L026 | Certificate of Analysis | 03/03/2025 | 031721.AP,031721.K7,031721.K2,031721.M1 |
Y15K050 | Certificate of Analysis | 19/11/2024 | 031721.AP,031721.K7,031721.K2,031721.M1 |
Y05K024 | Certificate of Analysis | 14/11/2024 | 031721.AP,031721.K7,031721.K2,031721.M1 |
W27K045 | Certificate of Analysis | 15/10/2024 | 031721.AP,031721.K7,031721.K2,031721.M1 |
T01K015 | Certificate of Analysis | 17/07/2024 | 031721.AP,031721.K7,031721.K2,031721.M1 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.