missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, anhydrous ≥ 99.8%, ChromAR™ for HPLC, Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
2.5 L
Description
- Ultraviolet Absorbance (1.00-cm cell vs. water): 0
- 210 nm ≤ 0.6
- 220 nm ≤ 0.3
- 230 nm ≤ 0.2
- 240 nm ≤ 0.05
- 260 nm ≤ 0.01
Specifications
Specifications
CAS | 67-56-1 |
Color | ≤10 (APHA) |
Quantity | 2.5 L |
Assay Percent Range | ≥ 99.8% |
Residue after Evaporation | ≤ 0.0005% |
Molecular Formula | CH4O |
Titratable Acid | ≤ 0.0005 meq/g |
Titratable Base | ≤ 0.0005 meq/g |
MDL Number | MFCD00004595 |
UN Number | 1230 |
Show More |
Safety and Handling



Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction