Learn More
Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
38.89 € - 67.13 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow More |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304807
|
Thermo Scientific Alfa Aesar
041838.AK |
250 mL |
38.89 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314807
|
Thermo Scientific Alfa Aesar
041838.K2 |
1 L |
67.13 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow More | |
CHEBI:17790 | |
CO |
Specifications
°C to 98°C | |
0.791 g/mL | |
11°C (52°F) | |
1.329 | |
CH4O | |
1098229 | |
Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone. | |
CO | |
32.04 | |
CHEBI:17790 | |
Methanol, anhydrous |
67-56-1 | |
64.7°C | |
Liquid | |
99.9% | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370-H372
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 200-659-6
missing translation for 'rtecsNumber' : PC1400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : methanol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.