missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, BAKER ANALYZED™ ACS, J.T. Baker™
Exceeds ACS Specifications
50.00 € - 102.00 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | Hydroxy methane , Methyl alcohol , Carbinol , Wood alcohol |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10281001
|
JT Baker
8045.1000 |
1 L | Glass Bottle |
50.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10179100
|
JT Baker
8045.2500 |
2.5 L | Glass Bottle |
102.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15538464
|
JT Baker
8045.2500PE |
2.5 L | Plastic Bottle |
78.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |
CH4O | |
MFCD00004595 | |
Hydroxy methane , Methyl alcohol , Carbinol , Wood alcohol | |
CO |
Specifications
-98°C | |
Max. 10 (APHA) | |
0.7918 g/cm³ (20°C) | |
11°C | |
max. 0.001 % | |
MFCD00004595 | |
Hydroxy methane , Methyl alcohol , Carbinol , Wood alcohol | |
CO | |
32.04 |
67-56-1 | |
Clear | |
64.6°C (1013 hPa) | |
min. 99.8 % (by GC, corrected for water) | |
CH4O | |
max. 0.05 % | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |
Safety and Handling
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title