missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, BioReagent, ≥99.93%, For Protein Sequencing, Honeywell Burdick & Jackson™
BioReagent, suitable for protein sequencing,≥99.93%
83.50 € - 124.99 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15615670
|
Honeywell-Burdick and Jackson
494437-1L |
1 L | Glass Bottle |
83.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15625670
|
Honeywell-Burdick and Jackson
494437-2L |
2 L | Glass Bottle |
124.99 €
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98°C | |
0.791g/mL at 25°C | |
0.36 | |
725°F | |
Liquid | |
CH3OH | |
410mmHg (50°C), 97.68mmHg (20°C) | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
Methanol |
67-56-1 | |
65°C | |
9°C (48.2°F) | |
Characteristic | |
CH4O | |
MFCD00004595 | |
UN1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.93% |
Safety and Handling
P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
missing translation for 'dotInformation' : class 3 - PG 2 - Methanol
missing translation for 'rtecsNumber' : 200-659-6