missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Capillary GC Grade; ≥99.9%, Honeywell Riedel-de Haën™
Capillary GC grade,≥99.9%
199.87 € - 648.73 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15658400
|
Honeywell-Chromasolv
414719-4L |
4 L | Glass Bottle |
199.87 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15625460
|
Honeywell-Chromasolv
414719-4X4L |
4 x 4 L | Glass Bottle |
648.73 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For GC headspace applications
- Meets USP, Ph. Eur. and ICH requirements
- GC headspace application tested
- Packed under inert gas for longer shelf life
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-95°C | |
Colorless | |
64.7°C | |
9°C (48.2°F) | |
Characteristic | |
n20/D 1.329 | |
CH3OH | |
97.68mmHg (20°C), 410mmHg (50°C) | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
CHROMASOLV™ Capillary GC |
67-56-1 | |
0.791g/mL at 25°C, 0.791g/mL at 25°C | |
0.36 | |
725°F | |
Liquid | |
CH4O | |
MFCD00004595 | |
UN1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.9% | |
Methanol |
Safety and Handling
P210-P260-P280-P301 + P310-P311
missing translation for 'dotInformation' : class 3 - PG 2 - Methanol
missing translation for 'rtecsNumber' : 200-659-6