missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Methanol CHROMASOLV™

Product Code. 15975480
Click to view available options
Quantity:
7 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Methanol CHROMASOLV™ for pesticide residue analysis, ≥99.9%.

  • For gas chromatography (GC).
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC.
  • Non-volatile matter and water content are specified for each solvent.

Specifications

pH Not applicable
Melting Point -98°C
CAS 67-56-1
Color Colorless
Density 0.79 g/cm3 (20°C)
Boiling Point 64°C to 65°C (1.013 hPa)
Quantity 7 L
Flash Point 11°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV™
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
Vapor Pressure 128 hPa (20°C)
UN Number UN1230
Viscosity 0.55 mPa.s (25°C)
Beilstein 1098229
Synonym Methyl alcohol
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Percent Purity ≥99.90%
Grade GC
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.