missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d{4}, 99.8%(Isotopic), Thermo Scientific Chemicals

42.91 € - 368.84 €

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387958
View Documents
Thermo Scientific Alfa Aesar
036501.03
1 g
42.91 €
1g
Please to purchase this item. Need a web account? Register with us today!
11377958
View Documents
Thermo Scientific Alfa Aesar
036501.06
5 g
138.69 €
5g
Please to purchase this item. Need a web account? Register with us today!
11397958
View Documents
Thermo Scientific Alfa Aesar
036501.14
25 g
368.84 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Methyl alcohol-d4 is the labeled analog of methyl alcohol, a common laboratory solvent. It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

811-98-3
36.066
OKKJLVBELUTLKV-MZCSYVLQSA-N
71568
CO
CH4O
MFCD00044637
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
trideuterio(deuteriooxy)methane

Specifications

Liquid
Mild
-98°C
CH4O
UN1230
It is miscible in water,ethanol,ether,acetone and chloroform.
CO
36.066
36.07
Methanol-d4, Isotopic
0.89 g/mL
811-98-3
64.7°C
MFCD00044637
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
OKKJLVBELUTLKV-MZCSYVLQSA-N
trideuterio(deuteriooxy)methane
71568
99.8%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H310-H330-H370
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331-H370

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL

missing translation for 'einecsNumber' : 212-378-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.