missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10051084
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 0.8880g/mL
Deuterium (D) 99.75% min.
Flash Point 12°C
Refractive Index 1.3262
Water 0.03% max.
Quantity 100 mL
CAS 811-98-3
Melting Point -99.0°C
Boiling Point 65.0°C
Molecular Formula CH4O
Linear Formula CD3OD
MDL Number MFCD00044637
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES CO
IUPAC Name trideuterio(deuteriooxy)methane
Molecular Weight (g/mol) 36.066
PubChem CID 71568
Formula Weight 36.06
Grade NMR
Chemical Name or Material Methanol-d4, For NMR, 99.8 atom% D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol-d4
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'