missing translation for 'onlineSavingsMsg'
Learn More

Methanol, for HPLC, Fisher Chemical™

Codice prodotto. 10674922
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle

CH4O, CAS Number-67-56-1, columbian spirit, methyl alcohol, colonial spirit, methyl hydroxide, methylol, carbinol, pyroxylic spirit, wood spirit, wood naphtha, wood alcohol, 2.5L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 67-56-1
Molecular Formula CH4O
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifica

Melting Point -98°C
Color Colorless
Boiling Point 64.7°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55cp at 20°C
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Molecular Weight (g/mol) 32.04
Formula Weight 32.04g/mol
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • Methanol
Avvertenza
  • Danger
Categoria di pericolo
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.
Informazioni supplementari
  • MIXTURE LIST-Contains : Methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'