missing translation for 'onlineSavingsMsg'
Learn More

Methanol, for HPLC, Gradient Grade, ACS, Honeywell Burdick & Jackson™

Product Code. 16695883
Click to view available options
1000 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point -98°C
CAS 67-56-1
Color Colorless
CAS Max % 1
Density 0.79 g/cm3 (20°C)
Boiling Point 64°C to 65°C (1013 hPa)
Quantity 1000 L
Flash Point 11°C
Molecular Formula CH4O
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Chemical Name or Material Methanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'