missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, LC-MS Ultra CHROMASOLV™, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS Ultra, tested for UHPLC-MS
83.98 € - 253.74 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15621400
|
Honeywell-Chromasolv
14262-1L |
1 L |
83.98 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15631400
|
Honeywell-Chromasolv
14262-2L |
2 L |
253.74 €
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
67-56-1 | |
0.36 | |
725°F | |
CH3OH | |
97.68mmHg (20°C), 410mmHg (50°C) | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
Methanol |
0.791g/mL at 25°C | |
9°C (48.2°F) | |
CH4O | |
MFCD00004595 | |
1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
LC-MS Ultra CHROMASOLV™ |
Safety and Handling
P210-P260-P280-P301 + P310-P311
H225-H301 + H311 + H331-H370
missing translation for 'einecsNumber' : 200-659-6
missing translation for 'rtecsNumber' : PC1400000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title