Learn More
MP Biomedicals™ Methionine/Choline Control Diet, Pellet
For the study of non-alcoholic steatohepatitis disease
Brand:  MP Biomedicals™ 0296044150
 View more versions of this product
Product Code. 11415375
  • 5500.00 € / 50kg
Explore more special offers
Description
Description
  • Methionine/choline control diet (MCCD) triggers the resolution of hepatic inflammatory and fibrotic reactions and hepatocyte apoptosis, suggesting that MCDD-induced steatohepatitis is also reversible
  • CD diet is used in Non-alcoholic steatohepatitis study
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Pellets
50 kg
Room Temperature