missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Methionine/Choline Deficient Diet, Pellet

Methionine/Choline Deficient Diet is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development.

Brand:  MP Biomedicals™ 0296043910

Product Code. 11435365

  • 1490.00 € / 10kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Non-alcoholic steatohepatitis(NASH) was induced by feeding a methionine/choline-deficient diet (MCD-diet) for up to 5 weeks to mice
  • NASH is a disease with a high incidence, difficult diagnosis, and as yet no effective treatment
  • So,MCD-diet is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Diet
Pellets
10 kg
Store at Room Temperature(15-30°C).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.