missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCRA Target Analytes, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 8041-A
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
2,3,4,5-Tetrachlorophenol | 4901-51-3 | 1000μg/mL |
2,3,4,6-Tetrachlorophenol | 58-90-2 | 1000μg/mL |
2,3,5,6-Tetrachlorophenol | 935-95-5 | 1000μg/mL |
2,4,5-Trichlorophenol | 95-95-4 | 1000μg/mL |
2,4,6-Trichlorophenol | 88-06-2 | 1000μg/mL |
2,4-Dichlorophenol | 120-83-2 | 1000μg/mL |
2,4-Dimethylphenol | 105-67-9 | 1000μg/mL |
2,4-Dinitrophenol | 51-28-5 | 1000μg/mL |
2,6-Dichlorophenol | 87-65-0 | 1000μg/mL |
2-Chlorophenol | 95-57-8 | 1000μg/mL |
2-Cyclohexyl-4,6-dinitrophenol | 131-89-5 | 1000μg/mL |
2-Methylphenol | 95-48-7 | 1000μg/mL |
2-Nitrophenol | 88-75-5 | 1000μg/mL |
3-Methylphenol | 108-39-4 | 1000μg/mL |
4,6-Dinitro-2-methylphenol | 534-52-1 | 1000μg/mL |
4-Chloro-3-methylphenol | 59-50-7 | 1000μg/mL |
4-Methylphenol | 106-44-5 | 1000μg/mL |
4-Nitrophenol | 100-02-7 | 1000μg/mL |
Dinoseb | 88-85-7 | 1000μg/mL |
Pentachlorophenol | 87-86-5 | 1000μg/mL |
Phenol | 108-95-2 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
RCRA Target Analytes | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
4-Chloro-3-methylphenol; 2-Chlorophenol; 2-Cyclohexyl-4,6-dinitrophenol; 2,4-Dichlorophenol; 2 |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For US EPA Method 8041 | |
Isopropanol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Phthalates Esters Mix
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol, 2,4,5-trichlorophenol, 2,6-dichlorophenol, dinoseb, 2,3,4,6-tetrachlorophenol
Product intended for laboratory use only