Learn More
Methoxytrimethylsilane, 97+%, Thermo Scientific Chemicals
35.88 € - 340.70 €
Chemical Identifiers
CAS | 1825-61-2 |
---|---|
Molecular Formula | C4H12OSi |
Molecular Weight (g/mol) | 104.22 |
MDL Number | MFCD00008344,MFCD03458977 |
InChI Key | POPACFLNWGUDSR-UHFFFAOYSA-N |
Synonym | trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn |
PubChem CID | 15771 |
IUPAC Name | methoxy(trimethyl)silane |
SMILES | CO[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308988
|
Thermo Scientific Alfa Aesar
042464.09 |
10 g |
35.88 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388978
|
Thermo Scientific Alfa Aesar
042464.18 |
50 g |
132.66 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398978
|
Thermo Scientific Alfa Aesar
042464.30 |
250 g |
340.70 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methoxytrimethylsilane plays an important role in the synthesis of organic compounds and used as water disgusting. It is used in the preparation of alfa-lithioalkoxysilanes by treating with tert-butyllithium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1825-61-2 | |
104.22 | |
POPACFLNWGUDSR-UHFFFAOYSA-N | |
15771 | |
CO[Si](C)(C)C |
C4H12OSi | |
MFCD00008344,MFCD03458977 | |
trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn | |
methoxy(trimethyl)silane |
Specifications
57°C to 58°C | |
Liquid | |
MFCD00008344,MFCD03458977 | |
−11°C (−12°F) | |
Miscible with cyclopentane. | |
CO[Si](C)(C)C | |
104.22 | |
104.22 | |
Mild | |
Moisture sensitive | |
Methoxytrimethylsilane |
1825-61-2 | |
C4H12OSi | |
UN1993 | |
trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn | |
POPACFLNWGUDSR-UHFFFAOYSA-N | |
methoxy(trimethyl)silane | |
15771 | |
≥97% | |
1.3678 | |
0.756 g/mL |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 217-369-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.