missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-aminocyclobutanecarboxylate hydrochloride, 95%, Thermo Scientific™

Product Code. 15459086
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15459086

Brand: Thermo Scientific Alfa Aesar H64701.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as a primary and secondary intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92398-47-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD01318265
InChI Key SGXOPTPGZBLQAY-UHFFFAOYSA-N
Synonym methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester
PubChem CID 1502033
IUPAC Name methyl 1-aminocyclobutane-1-carboxylate
SMILES COC(=O)C1(N)CCC1

Specifications

Quantity 250 mg
Beilstein 5615121
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 165.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 1-aminocyclobutanecarboxylate hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.