missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(acetylamino)-3-nitrobenzoate, 97%, Thermo Scientific™

Product Code. 10581482
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 95067-27-9
Molecular Formula C10H10N2O5
Molecular Weight (g/mol) 238.20
MDL Number MFCD00157613
InChI Key PHFRWNCHBPVTJN-UHFFFAOYSA-N
Synonym methyl 2-acetylamino-3-nitrobenzoate, methyl 2-acetamido-3-nitro-benzoate, 2-acetylamino-3-nitro-benzoic acid methyl ester, 2-acetamido-3-nitrobenzoic acid methyl ester, methyl-2-acetylamino-3-nitrobenzoate, methyl2-acetylamino-3-nitrobenzoate, 2-acetylamino-3-nitrobenzoic acid methyl ester
PubChem CID 2782278
IUPAC Name methyl 2-acetamido-3-nitrobenzoate
SMILES COC(=O)C1=C(NC(C)=O)C(=CC=C1)[N+]([O-])=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.2
Percent Purity 97%
Chemical Name or Material Methyl 2-(acetylamino)-3-nitrobenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'