missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-hydroxy-4-methoxybenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 47°C to 51°C |
Boiling Point | 134°C (7 mmHg) |
Molecular Formula | C9H10O4 |
Quantity | 10 g |
Beilstein | 2691644 |
Synonym | methyl 4-methoxysalicylate, 2-hydroxy-4-methoxy-benzoic acid methyl ester, methyl-4-methoxysalicylate, benzoic acid, 2-hydroxy-4-methoxy-, methyl ester, 2-hydroxy-4-methoxybenzoic acid methyl ester, 4-methoxysalicylic acid methyl ester, acmc-2097hp, methyl 2-hydroxy-p-anisate, ksc494a7p, 2-methoxycarbonyl-5-methoxyphenol |
Solubility Information | Soluble in acetone. Insoluble in water. |
InChI Key | ZICRWXFGZCVTBZ-UHFFFAOYSA-N |
SMILES | COC1=CC(=C(C=C1)C(=O)OC)O |
IUPAC Name | methyl 2-hydroxy-4-methoxybenzoate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'