missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxo-2H-pyran-3-carboxylate, 98%

Product Code. 11435690
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11435690

Brand: Thermo Scientific Alfa Aesar H25932.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25991-27-9 | C7H6O4 | 154.121 g/mol

Methyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25991-27-9
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00075443
InChI Key GJVZWOMUTYNUCE-UHFFFAOYSA-N
Synonym methyl 2-oxo-2h-pyran-3-carboxylate, 3-carbomethoxy-2-pyrone, methyl 2-pyrone-3-carboxylate, 2-oxo-2h-pyran-3-carboxylic acid methyl ester, 2h-pyran-3-carboxylic acid, 2-oxo-, methyl ester, methoxycarbonyl pyranone, acmc-1clg5, ksc496k6b
PubChem CID 117704
IUPAC Name methyl 2-oxopyran-3-carboxylate
SMILES COC(=O)C1=CC=COC1=O

Specifications

Melting Point 74°C to 77°C
Boiling Point 146°C to 148°C (0.75 mmHg)
Quantity 1 g
Beilstein 1424749
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material Methyl 2-oxo-2H-pyran-3-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.