missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-bromoethoxy)benzoate, 97+%, Thermo Scientific™

Product Code. 15429127
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 3-(2-bromoethoxy)benzoate used in pharmaceutical research.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59516-96-0
Molecular Formula C10H11BrO3
Molecular Weight (g/mol) 259.10
MDL Number MFCD06204041
InChI Key CGGLKWTURIUCMN-UHFFFAOYSA-N
Synonym methyl 3-2-bromoethoxy benzoate, methyl 3-2-bromoethoxy benzenecarboxylate, benzoic acid,3-2-bromoethoxy-, methyl ester, methyl 3-2-bromoethoxy-benzoate, methyl 3-2-bromoethyloxy benzoate, 1-bromo-2-3'-carbomethoxyphenoxy ethane, 3-2-bromoethoxy benzoic acid methyl ester, 3-2-bromo-ethoxy-benzoic acid methyl ester
PubChem CID 10491483
IUPAC Name methyl 3-(2-bromoethoxy)benzoate
SMILES COC(=O)C1=CC(OCCBr)=CC=C1

Specifications

Quantity 250 mg
Formula Weight 259.1
Percent Purity ≥97%
Chemical Name or Material Methyl 3-(2-bromoethoxy)benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'