Learn More
Methyl 3,4-dihydroxybenzoate, MedChemExpress
MedChemExpress Methyl 3,4-dihydroxybenzoate (Protocatechuic acid methyl ester; Methyl protocatechuate) is a major metabolite of antioxidant polyphenols found in green tea. Antioxidant and anti-inflammatory effect.
Brand: Medchem Express HY-Z0548
This item is not returnable.
View return policy
Description
Methyl 3,4-dihydroxybenzoate (Protocatechuic acid methyl esterSpecifications
Methyl 3,4-dihydroxybenzoate | |
Off-White | |
C8H8O4 | |
Protocatechuic acid methyl ester Methyl protocatechuate | |
O=C(OC)C1=CC=C(O)C(O)=C1 | |
168.15 | |
Research | |
Cancer-programmed cell death |
2150-43-8 | |
Research | |
50 mg | |
DMSO : ≥ 100 mg/mL (594.71 mM) | |
168.15 | |
99.26% | |
Powder |
Research purposes only