Learn More
Methyl 3-aminothiophene-2-carboxylate, 98+%, Thermo Scientific Chemicals
Description
Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Specifications
Specifications
Melting Point | 63°C to 67°C |
Boiling Point | 100°C to 102°C (0.1 mmHg) |
Quantity | 25 g |
Beilstein | 1365614 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 157.19 |
Percent Purity | ≥98% |
Chemical Name or Material | Methyl 3-aminothiophene-2-carboxylate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0446737 | Certificate of Analysis | 07/12/2022 | A12802.06,A12802.14,A12802.22 |
10237973 | Certificate of Analysis | 12/04/2022 | A12802.06,A12802.14,A12802.22 |
10238676 | Certificate of Analysis | 12/04/2022 | A12802.06,A12802.14,A12802.22 |
USLY001091 | Certificate of Analysis | 22/11/2021 | A12802.06,A12802.14,A12802.22 |
F5980A | Certificate of Analysis | 22/11/2021 | A12802.06,A12802.14,A12802.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.