missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-fluorophenylacetate, 98%, Thermo Scientific Chemicals

Product Code. 10469242
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64123-77-9
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD03093288
InChI Key OXYLCGRXCQARQV-UHFFFAOYSA-N
Synonym methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
PubChem CID 2734108
IUPAC Name methyl 2-(3-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=CC(F)=C1

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2CO2H
Refractive Index 1.4870 to 1.489
Quantity 5 g
Formula Weight 168.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-fluorophenylacetate, 98%
Show More Show Less
Product Identifier
  • Methyl 3-fluorophenylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'