missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-isocyanatobenzoate, 97%, Maybridge™

Product Code. p-4813550
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 10075752

Brand: Thermo Scientific Maybridge AC31920DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 41221-47-0
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00037077
InChI Key WBGWGERFPSYHDT-UHFFFAOYSA-N
Synonym 3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate
PubChem CID 2733366
IUPAC Name methyl 3-isocyanatobenzoate
SMILES COC(=O)C1=CC=CC(=C1)N=C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 177.16
Chemical Name or Material Methyl 3-isocyanatobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.