missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11352846
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5368-81-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017193
InChI Key DUKYPQBGYRJVAN-UHFFFAOYSA-N
Synonym methyl m-anisate, methyl m-methoxybenzoate, 3-methoxybenzoic acid methyl ester, m-anisic acid, methyl ester, benzoic acid, 3-methoxy-, methyl ester, 3-methoxybenoic acid methyl ester, m-methoxybenzoic acid methyl ester, mehtyl 3-methoxybenzoate, acmc-1asq5, m-anisic acid methyl ester
PubChem CID 79332
IUPAC Name methyl 3-methoxybenzoate
SMILES COC(=O)C1=CC=CC(OC)=C1

Specifications

Density 1.15
Boiling Point 238°C
Flash Point >110°C (230°F)
Refractive Index 1.527
Quantity 10 g
Beilstein 2208567
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material Methyl 3-methoxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'