missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™

71.25 € - 267.33 €

Chemical Identifiers

CAS 93618-66-7
Molecular Formula C11H9F3O3
Molecular Weight (g/mol) 246.19
MDL Number MFCD00216522
InChI Key RPRMYRPHNDGZOY-UHFFFAOYSA-N
Synonym methyl 3-oxo-3-3-trifluoromethyl phenyl propanoate, methyl 3-trifluoromethyl benzoylacetate, methyl 3-trifluoromethyl benzoyl acetate, 3-3-trifluoromethylphenyl-3-oxo-propionic acid methyl ester, 3-oxo-3-3-trifluoromethyl-phenyl-propionic acid methyl ester, pubchem2695, methyl 3-trifluoromethylbenzoylacetate, methyl 3-oxo-3-3-trifluoromethylphenyl propanoate, 3-oxo-3-3-trifluoromethylphenyl propionic acid methyl ester
PubChem CID 735880
IUPAC Name methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate
SMILES COC(=O)CC(=O)C1=CC=CC(=C1)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10341551
View Documents
Thermo Scientific Acros
425450025
2.5 g Glass bottle
71.25 €
2.50g
Please to purchase this item. Need a web account? Register with us today!
10731715
View Documents
Thermo Scientific Acros
425450100
10 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

93618-66-7
246.19
RPRMYRPHNDGZOY-UHFFFAOYSA-N
735880
COC(=O)CC(=O)C1=CC=CC(=C1)C(F)(F)F
C11H9F3O3
MFCD00216522
methyl 3-oxo-3-3-trifluoromethyl phenyl propanoate, methyl 3-trifluoromethyl benzoylacetate, methyl 3-trifluoromethyl benzoyl acetate, 3-3-trifluoromethylphenyl-3-oxo-propionic acid methyl ester, 3-oxo-3-3-trifluoromethyl-phenyl-propionic acid methyl ester, pubchem2695, methyl 3-trifluoromethylbenzoylacetate, methyl 3-oxo-3-3-trifluoromethylphenyl propanoate, 3-oxo-3-3-trifluoromethylphenyl propionic acid methyl ester
methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate

Specifications

93618-66-7
94°C to 96°C (0.2 mmHg)
Authentic
C11H9F3O3
MFCD00216522
Solubility in water: neglegible in water
COC(=O)CC(=O)C1=CC=CC(=C1)C(F)(F)F
246.19
246.18
Liquid
Yellow
>200°C
94% min. (HPLC)
1.4864 to 1.4884
methyl 3-oxo-3-3-trifluoromethyl phenyl propanoate, methyl 3-trifluoromethyl benzoylacetate, methyl 3-trifluoromethyl benzoyl acetate, 3-3-trifluoromethylphenyl-3-oxo-propionic acid methyl ester, 3-oxo-3-3-trifluoromethyl-phenyl-propionic acid methyl ester, pubchem2695, methyl 3-trifluoromethylbenzoylacetate, methyl 3-oxo-3-3-trifluoromethylphenyl propanoate, 3-oxo-3-3-trifluoromethylphenyl propionic acid methyl ester
RPRMYRPHNDGZOY-UHFFFAOYSA-N
methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate
735880
95%
Methyl 3-trifluoromethylbenzoylacetate
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl 3-trifluoromethylbenzoylacetate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.