missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorocarbonylbenzoate, 90%, tech., Thermo Scientific Chemicals

Product Code. 10472985
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7377-26-6
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.61
MDL Number MFCD00059486
InChI Key CVXXHXPNTZBZEL-UHFFFAOYSA-N
Synonym methyl 4-chlorocarbonyl benzoate, methyl 4-chlorocarbonylbenzoate, methyl 4-chloroformyl benzoate, terephthalic acid monomethyl ester chloride, methyl 4-carbonochloridoyl benzoate, methyl terephthaloyl chloride, benzoic acid, 4-chlorocarbonyl-, methyl ester, 4-chlorocarbonyl-benzoic acid methyl ester, 4-carbomethoxybenzoylchloride, 4-chlorocarbonylbenzoic acid methyl ester
PubChem CID 81850
IUPAC Name methyl 4-carbonochloridoylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point 50°C to 59°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula H3COOCC6H4COCl
Quantity 5 g
Solubility Information Solubility in water: hydrolysis
Formula Weight 198.61
Percent Purity 90%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-chlorocarbonylbenzoate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl 4-chlorocarbonylbenzoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'