Learn More
Methyl 4-chlorocarbonylbenzoate, 98%, Thermo Scientific Chemicals
Description
4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.
Solubility
Hydrolyzes in water.
Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
Specifications
Specifications
Melting Point | 52°C to 56°C |
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 198.6 |
Percent Purity | ≥98% |
Chemical Name or Material | Methyl 4-chlorocarbonylbenzoate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.