missing translation for 'onlineSavingsMsg'
Learn More
Methyl 4-ethynylbenzoate, 97%, Thermo Scientific Chemicals Product Code.: 15460208

Methyl 4-ethynylbenzoate, 97%, Thermo Scientific Chemicals

Product Code. 15460208
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15460208

Brand: Thermo Scientific Alfa Aesar H53419.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.16 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-86-4
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00168820
InChI Key JPGRSTBIEYGVNO-UHFFFAOYSA-N
Synonym 4-ethynylbenzoic acid methyl ester, 4-ethynyl-benzoic acid methyl ester, benzoic acid, 4-ethynyl-, methyl ester, methyl4-ethynylbenzoate, 4-ethynylbenzoic acid methylester, 4-methoxycarbonyl phenylacetylene, benzoic acid,4-ethynyl-, methyl ester, methyl 4-ethynyl-benzoate, 4-ethinylbenzoic acid methyl ester, 4-ethynyl benzoic acid methylester
PubChem CID 640163
IUPAC Name methyl 4-ethynylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C#C

Specifications

Melting Point 92°C to 94°C
Quantity 5 g
Solubility Information Sparingly soluble in water (0.16g/L at 25°C).
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material Methyl 4-ethynylbenzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN20A053 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53419.06,H53419.03
Lot NumberN27D054 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53419.06,H53419.03
Lot NumberA17X061 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53419.06,H53419.03
Lot NumberA11Y007 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53419.06,H53419.03
Lot NumberF07Y006 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53419.06,H53419.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4-ethynylbenzoate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.