missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals

Product Code. 11489753
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-50-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007350
InChI Key YOJAHJGBFDPSDI-UHFFFAOYSA-N
Synonym methyl p-nitrobenzoate, benzoic acid, 4-nitro-, methyl ester, p-nitrobenzoic acid methyl ester, methyl-p-nitrobenzoate, methyl-4-nitrobenzoate, benzoic acid, p-nitro-, methyl ester, p-carbomethoxynitrobenzene, unii-nmn8cth864, nmn8cth864, 4-nitrobenzoic acid methyl ester
PubChem CID 12083
IUPAC Name methyl 4-nitrobenzoate
SMILES COC(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 95°C to 97°C
Quantity 100 g
Beilstein 1912198
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material Methyl 4-nitrobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.