missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-oxotetrahydrothiophene-3-carboxylate, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 160.19 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | Methyl 4-oxotetrahydrothiophene-3-carboxylate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction