missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-oxotetrahydrothiophene-3-carboxylate, Technical Grade, Thermo Scientific™

Product Code. 10752293
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2689-68-1
Molecular Formula C6H8O3S
Molecular Weight (g/mol) 160.187
InChI Key LEAKUJFYXNILRB-UHFFFAOYSA-N
Synonym methyl 4-oxotetrahydrothiophene-3-carboxylate, 4-carbomethoxytetrahydro-3-thiophenone, 4-carbomethoxy-3-thiophanone, 3-thiophenecarboxylic acid, tetrahydro-4-oxo-, methyl ester, methyl tetrahydro-4-oxo-3-thiophenecarboxylate, methyl 3-oxotetrahydrothiophene-4-carboxylate, 4-carbomethoxytetrahydro-3-thi ophenone, methyl 4-oxo-tetrahydrothiophene-3-carboxylate, methyl 4-oxo-2,3,5-trihydrothiophene-3-carboxylate, methyl tetrahydro-4-oxo-3-thenoate
PubChem CID 98424
IUPAC Name methyl 4-oxothiolane-3-carboxylate
SMILES COC(=O)C1CSCC1=O

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 160.19
Percent Purity Tech.
Grade Technical
Chemical Name or Material Methyl 4-oxotetrahydrothiophene-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.